MilliporeSigma
  • Benefit of neoadjuvant chemotherapy for Siewert type II esophagogastric junction adenocarcinoma.
Benefit of neoadjuvant chemotherapy for Siewert type II esophagogastric junction adenocarcinoma.
Anticancer research (2015-01-01)
Kei Hosoda, Keishi Yamashita, Natsuya Katada, Hiromitsu Moriya, Hiroaki Mieno, Shinichi Sakuramoto, Shiro Kikuchi, Masahiko Watanabe
ABSTRACT
Our objective was to clarify if preoperative chemotherapy was associated with improved survival in Japanese patients with Siewert type II adenocarcinoma of the esophagogastric junction. We retrospectively reviewed the medical records of 86 patients with Siewert type II adenocarcinoma who underwent R0 resection at the Kitasato University between 1997 and 2013. Cox regression analysis using a backward stepwise selection method was performed to identify independent prognostic factors for relapse-free survival (RFS). The median age was 67 years. The male:female ratio was 74:12. Right thoracic, left thoracic and transhiatal approaches were performed in 10, 10 and 66 patients, respectively, and perioperative transfusion in 16 patients. Preoperative chemotherapy was administered to 19 patients; out of these, 13 received chemotherapy using the DCS regimen (docetaxel 40 mg/m(2), day 1; cisplatin 60 mg/m(2), day 1; S-1 80-120 mg/body, days 1-14; every 28 days). A median of three cycles of preoperative DCS chemotherapy were used. Histological responses of 1b, 2, 3 and unknown grades were obtained in three, three, four and three patients, respectively. The 5-year RFS rate was 55%, and the median follow-up period was 36 months. Cox regression analysis regarding RFS identified (y)pN1-3 [hazard ratio (HR)=4.44; 95% confidence interval (CI)=1.98-11.27], performance of perioperative transfusion (HR=4.71; 95% CI=1.69-11.88) and no preoperative chemotherapy (HR=3.75; 95% CI=1.22-14.26) as significant and independent indicators of poor prognosis. Preoperative chemotherapy using DCS is potentially beneficial for Japanese patients with Siewert type II adenocarcinoma. Further prospective clinical studies are required to confirm our findings.
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Anhydrous Docetaxel, European Pharmacopoeia (EP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline