MilliporeSigma
  • Organocatalytic enantioselective addition of thiols to ketimines derived from isatins.
Organocatalytic enantioselective addition of thiols to ketimines derived from isatins.
Organic letters (2014-12-20)
Shuichi Nakamura, Shun Takahashi, Daisuke Nakane, Hideki Masuda
ABSTRACT
The first catalytic enantioselective addition of thiols to ketimines derived from isatins has been developed. Excellent yields and enantioselectivities were observed for the reaction of various ketimines and thiols using a cinchona alkaloid sulfonamide catalyst. Both enantiomers of products could be obtained by using pseudoenantiomeric chiral catalysts.
Sigma-Aldrich
Isatin, 97%