MilliporeSigma
  • Structure of the ATP-dependent oligomerization domain of N-ethylmaleimide sensitive factor complexed with ATP.
Structure of the ATP-dependent oligomerization domain of N-ethylmaleimide sensitive factor complexed with ATP.
Nature structural biology (1998-09-10)
R C Yu, P I Hanson, R Jahn, A T Brünger
ABSTRACT
N-ethylmaleimide-sensitive factor (NSF) is a hexameric ATPase which primes and/or dissociates SNARE complexes involved in intracellular fusion events. Each NSF protomer contains three domains: an N-terminal domain required for SNARE binding and two ATPase domains, termed D1 and D2, with D2 being required for oligomerization. We have determined the 1.9 A crystal structure of the D2 domain of NSF complexed with ATP using multi-wavelength anomalous dispersion phasing. D2 consists of a nucleotide binding subdomain with a Rossmann fold and a C-terminal subdomain, which is structurally unique among nucleotide binding proteins. There are interactions between the ATP moiety and both the neighboring D2 protomer and the C-terminal subdomain that may be important for ATP-dependent oligomerization. Of particular importance are three well-ordered and conserved lysine residues that form ionic interactions with the beta- and gamma-phosphates, one of which likely contributes to the low hydrolytic activity of D2.