MilliporeSigma
  • An efficient one-pot synthesis of coumarins mediated by propylphosphonic anhydride (T3P) via the Perkin condensation.
An efficient one-pot synthesis of coumarins mediated by propylphosphonic anhydride (T3P) via the Perkin condensation.
Tetrahedron Letters (2012)
Augustine JK, et al.