MilliporeSigma
  • An efficient catalytic method for the Friedlander annulation mediated by peptide coupling agent propylphosphonic anhydride (T3P?).
An efficient catalytic method for the Friedlander annulation mediated by peptide coupling agent propylphosphonic anhydride (T3P?).
Tetrahedron Letters (2011)
Augustine JK, et al.