MilliporeSigma
  • Rab8 binding to immune cell-specific adaptor LAX facilitates formation of trans-Golgi network-proximal CTLA-4 vesicles for surface expression.
Rab8 binding to immune cell-specific adaptor LAX facilitates formation of trans-Golgi network-proximal CTLA-4 vesicles for surface expression.
Molecular and cellular biology (2014-02-12)
Matthew C Banton, Kerry L Inder, Elke Valk, Christopher E Rudd, Helga Schneider
ABSTRACT
Despite playing a central role in tolerance, little is known regarding the mechanism by which intracellular CTLA-4 is shuttled from the trans-Golgi network to the surfaces of T cells. In this context, Ras-related GTPase Rab8 plays an important role in the intracellular transport, while we have previously shown that CTLA-4 binds to the immune cell adaptor TRIM in T cells. In this study, we demonstrate that CTLA-4 forms a multimeric complex comprised of TRIM and related LAX that in turn binds to GTP bound Rab8 for post-Golgi transport to the cell surface. LAX bound via its N terminus to active GTP-Rab8, as well as the cytoplasmic tail of CTLA-4. TRIM required LAX for binding to Rab8 in a complex. Wild-type LAX or its N terminus (residues 1 to 77) increased CTLA-4 surface expression, whereas small interfering RNAs of Rab8 or LAX or disruption of LAX/Rab8 binding reduced numbers of CTLA-4-containing vesicles and its coreceptor surface expression. LAX also promoted the polarization of CTLA-4 and the reorientation of the microtubule-organizing center to the site of T-cell receptor engagement. Our results identify a novel CTLA-4/TRIM/LAX/Rab8 effector complex in the transport of CTLA-4 to the surfaces of T cells.