MilliporeSigma
  • A comparative proteome analysis links tyrosine kinase 2 (Tyk2) to the regulation of cellular glucose and lipid metabolism in response to poly(I:C).
A comparative proteome analysis links tyrosine kinase 2 (Tyk2) to the regulation of cellular glucose and lipid metabolism in response to poly(I:C).
Journal of proteomics (2011-07-27)
Tom Grunert, Nicole R Leitner, Martina Marchetti-Deschmann, Ingrid Miller, Barbara Wallner, Marta Radwan, Claus Vogl, Thomas Kolbe, Dagmar Kratky, Manfred Gemeiner, Günter Allmaier, Mathias Müller, Birgit Strobl
ABSTRACT
Tyrosine kinase 2 (Tyk2) is an integral part of the Janus kinase-signal transducer and activator of transcription (JAK-STAT) pathway which relays intracellular signals of various cytokines. Tyk2 crucially contributes to host defense mechanisms against microbial pathogens and to tumor surveillance but also facilitates immune pathologies. Here we investigated the impact of Tyk2 on the macrophage proteome using the synthetic double-stranded RNA analog polyinosinic acid-polycytidylic acid (poly(I:C)) as a mimicry of viral infections. By means of 2D-DIGE in connection with PMF obtained by MALDI-MS and sequence tag determination by MS/MS we unambiguously identified eighteen protein spots corresponding to sixteen distinct proteins that are regulated by poly(I:C) and differentially expressed between wildtype (WT) and Tyk2-deficient macrophages. The majority of these proteins are functionally assigned to cellular immune responses and to metabolism. We show for selected metabolic enzymes, i.e. triosephosphate isomerase (TIM), ATP-citrate synthase (ACLY) and long-chain-fatty-acid-CoA ligase 4 (ACSL4), that Tyk2 affects protein expression transcriptionally and post-transcriptionally. We furthermore confirm the involvement of Tyk2 in the regulation of lipid and carbohydrate metabolism at the level of metabolites. Taken together, our results provide new evidence for important functions of Tyk2 at the molecular interface between innate immunity and cellular metabolism.