MilliporeSigma
HBXIP functions as a cofactor of survivin in apoptosis suppression.
The EMBO journal (2003-05-30)
Hiroyuki Marusawa, Shu-Ichi Matsuzawa, Kate Welsh, Hua Zou, Robert Armstrong, Ingo Tamm, John C Reed
ABSTRACT
Survivin is an anti-apoptotic protein that is overexpressed in most human cancers. We show that survivin forms complexes with a cellular protein, hepatitis B X-interacting protein (HBXIP), which was originally recognized for its association with the X protein of hepatitis B virus (HBX). Survivin-HBXIP complexes, but neither survivin nor HBXIP individually, bind pro-caspase-9, preventing its recruitment to Apaf1, and thereby selectively suppressing apoptosis initiated via the mitochondria/cytochrome c pathway. Viral HBX protein also interacts with the survivin- HBXIP complex and suppresses caspase activation in a survivin-dependent manner. Thus, HBXIP functions as a cofactor for survivin, and serves as a link between the cellular apoptosis machinery and a viral pathogen involved in hepatocellular carcinogenesis.