MilliporeSigma
  • Formal total syntheses of the (-)-salicylihalamides A and B from D-glucose and L-rhamnose.
Formal total syntheses of the (-)-salicylihalamides A and B from D-glucose and L-rhamnose.
The Journal of organic chemistry (2005-09-10)
Torsten Haack, KyoungLang Haack, Wibke E Diederich, Burchelle Blackman, Subho Roy, Srinivas Pusuluri, Gunda I Georg
ABSTRACT
[reaction: see text] Two formal total syntheses of the (-)-salicylihalamides, based on chiral pool approaches, are reported. D-glucose and L-rhamnose were used to prepare advanced intermediates 23 and 54, which can be converted in three or four steps, respectively, to the target compounds. The synthesis of 23 from a known D-glucose-derivative was accomplished in 12 steps and 17% overall yield, and the synthesis of 54 from a known L-rhamnose-derivative was done in nine steps and 6% overall yield. A key step in the synthesis was a ring-closing metathesis reaction to prepare the macrocyclic ring system. It was demonstrated that the phenolic protecting group was critical for inducing the preferential formation of the desired E isomer. It was further shown that the protecting group at the C13 hydroxyl group had no significant influence on the E:Z ratio during the ring-closing metathesis reaction.