MilliporeSigma
  • Solubility controlled electropolymerisation and study of the impact of regioregularity on the spectroelectrochemical properties of thin films of poly (3-octylthiophenes).
Solubility controlled electropolymerisation and study of the impact of regioregularity on the spectroelectrochemical properties of thin films of poly (3-octylthiophenes).
Electrochimica Acta (2014)
Jarosz T, et al.