MilliporeSigma
  • Genetic variability of prostaglandin E2 receptor subtype EP4 gene in aspirin-intolerant chronic urticaria.
Genetic variability of prostaglandin E2 receptor subtype EP4 gene in aspirin-intolerant chronic urticaria.
Journal of human genetics (2012-06-15)
Nami Shrestha Palikhe, Hye Jung Sin, Seung Hyun Kim, Hyun Jung Sin, Eui Kyung Hwang, Young Min Ye, Hae-Sim Park
ABSTRACT
Prostaglandin E2 receptor subtype EP4 (PTGER4) is one of the four subtypes of receptors for prostaglandin E2 (PGE2). Overproduction of cysteinyl leukotriene in mast cells may be related with suppression of PGE2 in patients with aspirin hypersensitivity. Considering the association of PTGER4 in mast cells, urticaria- and aspirin-related disease, we hypothesized the genetic variability of PTGER4 may be associated with aspirin-intolerant chronic urticaria (AICU). The case-control study was performed in 141 with AICU, 153 with aspirin-tolerant chronic urticaria (ATCU) and 174 with normal controls (NCs). PTGER4 promoter single-nucleotide polymorphism was genotyped using a primer extension method with the SNAPshot ddNTP primer extension kit. The functional variability of PTGER4 promoter polymorphism was carried out by dual-luciferase system and electrophoretic mobility shift assay (EMSA) in human mast cells (HMC-1). Furthermore, the effect of aspirin was performed for PTGER4 mRNA expression using real-time PCR, and PGE2 production was checked in HMC-1 cells using ELISA. AICU patients carrying GG genotype at -1254 G>A showed significantly higher frequency compared with NC (P=0.032). Similarly, the minor allele frequency, G allele was significantly higher in AICU compared with NC (P=0.031). In vitro functional study demonstrated that the -1254 G allele had lower luciferase activity (P<0.001) in HMC-1 cells. EMSA finding showed that PTGER4 -1254 G produced a specific band. Significantly decreased PTGER4 expression (P=0.008) and PGE2 production by aspirin exposure was confirmed in in vitro HMC cell line model (P=0.001). The PTGER4 -1254 G allele demonstrated a higher frequency in AICU patients and lower promoter activity with decreased expression of PTGER4 and contributes to the development of AICU.