MilliporeSigma
  • A History of the Production and Use of Carbon Tetrachloride, Tetrachloroethylene, Trichloroethylene and 1, 1, 1-Trichloroethane in the United States: Part 2--Trichloroethylene and 1, 1, 1-Trichloroethane.
A History of the Production and Use of Carbon Tetrachloride, Tetrachloroethylene, Trichloroethylene and 1, 1, 1-Trichloroethane in the United States: Part 2--Trichloroethylene and 1, 1, 1-Trichloroethane.
Environmental Forensics (2000)
Doherty RE.