MilliporeSigma
  • Phytochemical Composition, Antioxidant Activity, and Neuroprotective Effect of Terminalia chebula Retzius Extracts.
Phytochemical Composition, Antioxidant Activity, and Neuroprotective Effect of Terminalia chebula Retzius Extracts.
Evidence-based complementary and alternative medicine : eCAM (2011-07-15)
Chia Lin Chang, Che San Lin
ABSTRACT
The objectives of this study were to determine phytochemical compositions, chemiluminescence antioxidant activities, and neuroprotective effects on PC12 cells for water, methanol, and 95% ethanol extracts of the air-dried fruit of Terminalia chebula Retzius. The water extract afforded the greatest yield, and total phenolic and tannin content. The methanol extract yielded the greatest total triterpenoid content. Based on four chemiluminescence antioxidant assays, the three extracts showed various degrees of antioxidant activity. The methanol extract showed good antioxidant activity based on the horseradish peroxidase-luminol-hydrogen peroxide (H(2)O(2)) assay. The water extract appeared to have good antioxidant activities in cupric sulfate-Phen-Vc-H(2)O(2) and luminol-H(2)O(2) assays. Pyrogallol-luminol assay showed the 95% ethanol extract to have good antioxidant activity. The methanol and water extracts presented neuroprotective activities on H(2)O(2)-induced PC12 cell death at 0.5-5.0 μg/mL. Further investigations are necessary to verify these activities in vivo.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
Pricing and availability is not currently available.