MilliporeSigma
  • Heterogeneous reactivity of gaseous nitric acid on Al2O3, CaCO3, and atmospheric dust samples: A Knudsen cell study.
Heterogeneous reactivity of gaseous nitric acid on Al<SUB>2</SUB>O<SUB>3</SUB>, CaCO<SUB>3</SUB>, and atmospheric dust samples: A Knudsen cell study.
The Journal of Physical Chemistry A (2001)
Hanisch F and Crowley JN.