MilliporeSigma
  • Detection of carboxylic acid group in organic compounds with eosin, mercurochrome, and methylene blue.
Detection of carboxylic acid group in organic compounds with eosin, mercurochrome, and methylene blue.
Fresenius' Zeitschrift fur Analytische Chemie (1972)
Johar GS.