MilliporeSigma
  • Secretory carrier membrane protein SCAMP2 and phosphatidylinositol 4,5-bisphosphate interactions in the regulation of dense core vesicle exocytosis.
Secretory carrier membrane protein SCAMP2 and phosphatidylinositol 4,5-bisphosphate interactions in the regulation of dense core vesicle exocytosis.
Biochemistry (2007-08-24)
Haini Liao, Jeff Ellena, Lixia Liu, Gabor Szabo, David Cafiso, David Castle
ABSTRACT
Secretory carrier membrane protein 2 (SCAMP2) functions in late steps of membrane fusion in calcium-dependent granule exocytosis. A basic/hydrophobic peptide segment within SCAMP2 (SCAMP2 E: CWYRPIYKAFR) has been implicated in this function and shown to bind and sequester phosphatidylinositol 4,5-bisphosphate [PI(4,5)P2 or PIP2] within membranes through an electrostatic mechanism. We now show that alanine substitution of tryptophan W2 within SCAMP2 E substantially weakens peptide binding to negatively charged liposomes; other substitutions for arginine R4 and lysine K8 have only limited effects on binding. Electron paramagnetic resonance analysis of liposomes containing spin-labeled PIP2 shows that R4 but not K8 is critical for SCAMP E binding to PIP2. The interfacial locations of SCAMP E and its structural variants within lipid bicelles measured by oxygen enhancement of nuclear relaxation are all similar. Corresponding point mutations within full-length SCAMP2 (SC2-R204A, SC2-K208A, and SC2-W202A) have been analyzed for biological effects on dense core vesicle exocytosis in neuroendocrine PC12 cells. With the same level of overexpression, SC2-R204A but not SC2-K208A inhibited secretion of cotransfected human growth hormone and of noradrenalin. Inhibition by SC2-R204A was the same as or greater than previously observed for SC2-W202A. Analysis of noradrenalin secretion by amperometry showed that inhibitory mutants of SCAMP2 decrease the probability of fusion pore opening and the stability of initially opened but not yet expanded fusion pores. The strong correlation between SCAMP2 E interactions with PIP2 and inhibition of exocytosis, particularly by SC2-R204A, led us to propose that SCAMP2 interaction with PIP2 within the membrane interface regulates fusion pore formation during exocytosis.