MilliporeSigma
  • Ceramide kinase promotes Ca2+ signaling near IgG-opsonized targets and enhances phagolysosomal fusion in COS-1 cells.
Ceramide kinase promotes Ca2+ signaling near IgG-opsonized targets and enhances phagolysosomal fusion in COS-1 cells.
Journal of lipid research (2007-12-25)
Vania Hinkovska-Galcheva, Andrea Clark, Susan VanWay, Ji-Biao Huang, Miki Hiraoka, Akira Abe, Michael Borofsky, Robin G Kunkel, Thomas Shanley, James A Shayman, Frederick Lanni, Howard R Petty, Laurence A Boxer
ABSTRACT
Ceramide-1-phosphate (C1P) is a novel bioactive sphingolipid formed by the phosphorylation of ceramide catalyzed by ceramide kinase (CERK). In this study, we evaluated the mechanism by which increased C1P during phagocytosis enhances phagocytosis and phagolysosome formation in COS-1 cells expressing hCERK. Stable transfectants of COS-1 cells expressing FcgammaRIIA or both FcgammaRIIA/hCERK expression vectors were created. Cell fractionation studies demonstrated that hCERK and the transient receptor potential channel (TRPC-1) were enriched in caveolae fractions. Our data establish that both CERK and TRPC-1 localize to the caveolar microdomains during phagocytosis and that CERK also colocalizes with EIgG in FcgammaRIIA/hCERK-bearing COS-1 cells. Using high-speed fluorescence microscopy, FcgammaRIIA/hCERK transfected cells displayed Ca2+ sparks around the phagosome. In contrast, cells expressing FcgammaRIIA under identical conditions displayed little periphagosomal Ca2+ signaling. The enhanced Ca2+ signals were accompanied by enhanced phagolysosome formation. However, the addition of pharmacological reagents that inhibit store-operated channels (SOCs) reduced the phagocytic index and phagolysosomal fusion in hCERK transfected cells. The higher Ca2+ signal observed in hCERK transfected cells as well as the fact that CERK colocalized with EIgG during phagocytosis support our hypothesis that Ca2+ signaling is an important factor for increasing phagocytosis and is regulated by CERK in a manner that involves SOCs/TRPCs.
Sigma-Aldrich
Ceramide Kinase FLAG tag active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥40% (SDS-PAGE)