MilliporeSigma
  • Influence of non-essential amino acids on apoptotic and necrotic death of mouse hybridoma cells in batch cultures
Influence of non-essential amino acids on apoptotic and necrotic death of mouse hybridoma cells in batch cultures
Biotechnology Letters (1997)
Fassnacht D