MilliporeSigma
  • Direct measurement of Criegee intermediate (CH2OO) reactions with acetone, acetaldehyde, and hexafluoroacetone.
Direct measurement of Criegee intermediate (CH<SUB>2</SUB>OO) reactions with acetone, acetaldehyde, and hexafluoroacetone.
Physical Chemistry Chemical Physics (2012)
Taatjes CA, et al.