MilliporeSigma
  • Asymmetric Catalytic Alkynylation of Acetaldehyde: Application to the Synthesis of (+)-Tetrahydropyrenophorol.
Asymmetric Catalytic Alkynylation of Acetaldehyde: Application to the Synthesis of (+)-Tetrahydropyrenophorol.
Angewandte Chemie (International Edition in English) (2012)
Trost BM and Quintard A.