MilliporeSigma
  • One-pot cross double-Mannich reaction of acetaldehyde catalyzed by a binaphthyl-based amino sulfonamide.
One-pot cross double-Mannich reaction of acetaldehyde catalyzed by a binaphthyl-based amino sulfonamide.
Chemical Communications (Cambridge, England) (2013)
Kano T, et al.