MilliporeSigma
  • PRELI is a mitochondrial regulator of human primary T-helper cell apoptosis, STAT6, and Th2-cell differentiation.
PRELI is a mitochondrial regulator of human primary T-helper cell apoptosis, STAT6, and Th2-cell differentiation.
Blood (2008-10-24)
Johanna Tahvanainen, Teemu Kallonen, Hanna Lähteenmäki, Kaisa M Heiskanen, Jukka Westermarck, Kanury V S Rao, Riitta Lahesmaa
ABSTRACT
The identification of novel factors regulating human T helper (Th)-cell differentiation into functionally distinct Th1 and Th2 subsets is important for understanding the mechanisms behind human autoimmune and allergic diseases. We have identified a protein of relevant evolutionary and lymphoid interest (PRELI), a novel protein that induces oxidative stress and a mitochondrial apoptosis pathway in human primary Th cells. We also demonstrated that PRELI inhibits Th2-cell development and down-regulates signal transducer and activator of transcription 6 (STAT6), a key transcription factor driving Th2 differentiation. Our data suggest that calpain, an oxidative stress-induced cysteine protease, is involved in the PRELI-induced down-regulation of STAT6. Moreover, we observed that a strong T-cell receptor (TCR) stimulus induces expression of PRELI and inhibits Th2 development. Our results suggest that PRELI is involved in a mechanism wherein the strength of the TCR stimulus influences the polarization of Th cells. This study identifies PRELI as a novel factor influencing the human primary Th-cell death and differentiation.