MilliporeSigma
  • The Rates of Methoxyl Exchange of Camphor and Norcamphor Dimethyl Ketals in Methanol-d4.
The Rates of Methoxyl Exchange of Camphor and Norcamphor Dimethyl Ketals in Methanol-<I>d</I><SUB>4</SUB>.
Journal of the American Chemical Society (1966)
Traylor TG and Perrin CL.