MilliporeSigma
  • Determination of aldoses and ketoses by GC-MS using differential derivatisation.
Determination of aldoses and ketoses by GC-MS using differential derivatisation.
Phytochemical analysis : PCA (2006-12-06)
Fangting Ye, Xiaojun Yan, Jilin Xu, Haimin Chen
ABSTRACT
A method has been established by which to determine aldoses and ketoses in plant material simultaneously. Monosaccharides were extracted by sonication with 80% ethanol and sugar oximes formed by treatment of the resultant extract with hydroxylamine and pyridine at 90 degrees C. After reaction, one aliquot of the product was derivatised with acetic anhydride at 90 degrees C, whilst a second aliquot was silylated with HMDS and TMCS at 80 degrees C. Both reaction mixtures were analysed by GC-MS in the SIM mode. Quantivation was linear within the range 1-4 microg/mL and the detection limit for monosaccharides was 5-25 ng/mL. The absolute recoveries were between 73.0 and 90.2% and the RSDs were 3.1-10.0%. This method was applied to analyse the free monosaccharides in Lyceum barbarum L.; eight monosaccharides were present in amounts between 0.26 and 368.65 microg/mg.