MilliporeSigma
  • Preliminary structural characterization of human SOUL, a haem-binding protein.
Preliminary structural characterization of human SOUL, a haem-binding protein.
Acta crystallographica. Section F, Structural biology and crystallization communications (2009-07-04)
Filipe Freire, Maria João Romão, Anjos L Macedo, Susana S Aveiro, Brian J Goodfellow, Ana Luísa Carvalho
ABSTRACT
Human SOUL (hSOUL) is a 23 kDa haem-binding protein that was first identified as the PP(23) protein isolated from human full-term placentas. Here, the overexpression, purification and crystallization of hSOUL are reported. The crystals belonged to space group P6(4)22, with unit-cell parameters a = b = 145, c = 60 A and one protein molecule in the asymmetric unit. X-ray diffraction data were collected to 3.5 A resolution at the ESRF. A preliminary model of the three-dimensional structure of hSOUL was obtained by molecular replacement using the structures of murine p22HBP (PDB codes 2gov and 2hva), obtained by solution NMR, as search models.