MilliporeSigma
  • Molecular and biochemical characterization of Paragonimus westermani tyrosinase.
Molecular and biochemical characterization of Paragonimus westermani tyrosinase.
Parasitology (2015-01-27)
Y-A Bae, S-H Kim, C-S Ahn, J-G Kim, Y Kong
ABSTRACT
Trematode tyrosinases (TYRs) play a major role in the tanning process during eggshell formation. We investigated the molecular and biochemical features of Paragonimus westermani TYR (PwTYR). The PwTYR cDNA was composed of 1568-bp encompassing a 1422-bp-long open reading frame (474-amino acid polypeptide). A strong phylogenetic relationship with Platyhelminthes and Deuterostomian orthologues was evident. The recombinant PwTYR expressed in prokaryotic cells promptly oxidized diphenol substrates, with a preferential affinity toward ortho-positioned hydroxyl groups. It demonstrated fairly weak activity for monophenol compounds. Diphenol oxidase activity was augmented with an increase of pH from 5.0 to 8.0, while monophenol oxidase activity was highest at an acidic pH and gradually decreased as pH increased. Transcription profile of PwTYR was temporally upregulated along with worm development. PwTYR was specifically localized in vitellocytes and eggs. The results suggested that conversion of tyrosine to L-dihydroxyphenylalanine by PwTYR monophenol oxidase activity might be rate-limiting step during the sclerotization process of P. westermani eggs. The pH-dependent pattern of monophenol and diphenol oxidase activity further proposes that the initial hydroxylation might slowly but steadily progress in acidic secreted vesicles of vitellocytes and the second oxidation process might be rapidly accelerated by neural or weak alkaline pH environments within the ootype.
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Hydroquinone, JIS special grade, ≥99.0%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
USP
Hydroquinone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydroquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nitrilotriacetic acid, BioUltra, ≥99.0% (T)
Supelco
Nitrilotriacetic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Levodopa, European Pharmacopoeia (EP) Reference Standard
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material