MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Polyunsaturated fatty acids inhibit stimulated coupling between the ER Ca(2+) sensor STIM1 and the Ca(2+) channel protein Orai1 in a process that correlates with inhibition of stimulated STIM1 oligomerization.
Polyunsaturated fatty acids inhibit stimulated coupling between the ER Ca(2+) sensor STIM1 and the Ca(2+) channel protein Orai1 in a process that correlates with inhibition of stimulated STIM1 oligomerization.
Biochimica et biophysica acta (2014-04-29)
David Holowka, Marek K Korzeniowski, Kirsten L Bryant, Barbara Baird
ABSTRACT
Polyunsaturated fatty acids (PUFAs) have been found to be effective inhibitors of cell signaling in numerous contexts, and we find that acute addition of micromolar PUFAs such as linoleic acid effectively inhibit of Ca(2+) responses in mast cells stimulated by antigen-mediated crosslinking of FcεRI or by the SERCA pump inhibitor, thapsigargin. In contrast, the saturated fatty acid, stearic acid, with the same carbon chain length as linoleic acid does not inhibit these responses. Consistent with this inhibition of store-operated Ca(2+) entry (SOCE), linoleic acid inhibits antigen-stimulated granule exocytosis to a similar extent. Using the fluorescently labeled plasma membrane Ca(2+) channel protein, AcGFP-Orai1, together with the labeled ER Ca(2+) sensor protein, STIM1-mRFP, we monitor stimulated coupling of these proteins that is essential for SOCE with a novel spectrofluorimetric resonance energy transfer method. We find effective inhibition of this stimulated coupling by linoleic acid that accounts for the inhibition of SOCE. Moreover, we find that linoleic acid induces some STIM1-STIM1 association, while inhibiting stimulated STIM1 oligomerization that precedes STIM1-Orai1 coupling. We hypothesize that linoleic acid and related PUFAs inhibit STIM1-Orai1 coupling by a mechanism that involves perturbation of ER membrane structure, possibly by disrupting electrostatic interactions important in STIM1 oligomerization. Thisarticle is part of a Special Issue entitled Tools to study lipid functions.
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Stearic acid, SAJ first grade, ≥90.0%, powder
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Stearic acid, European Pharmacopoeia (EP) Reference Standard
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, analytical standard