MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Selectivity of natural, synthetic and environmental estrogens for zebrafish estrogen receptors.
Selectivity of natural, synthetic and environmental estrogens for zebrafish estrogen receptors.
Toxicology and applied pharmacology (2014-08-12)
Caroline Pinto, Marina Grimaldi, Abdelhay Boulahtouf, Farzad Pakdel, François Brion, Sélim Aït-Aïssa, Vincent Cavaillès, William Bourguet, Jan-Ake Gustafsson, Maria Bondesson, Patrick Balaguer
ABSTRACT
Zebrafish, Danio rerio, is increasingly used as an animal model to study the effects of pharmaceuticals and environmental estrogens. As most of these estrogens have only been tested on human estrogen receptors (ERs), it is necessary to measure their effects on zebrafish ERs. In humans there are two distinct nuclear ERs (hERα and hERβ), whereas the zebrafish genome encodes three ERs, zfERα and two zfERβs (zfERβ1 and zfERβ2). In this study, we established HeLa-based reporter cell lines stably expressing each of the three zfERs. We first reported that estrogens more efficiently activate the zfERs at 28°C as compared to 37°C, thus reflecting the physiological temperature of zebrafish in wildlife. We then showed significant differences in the ability of agonist and antagonist estrogens to modulate activation of the three zfER isotypes in comparison to hERs. Environmental compounds (bisphenol A, alkylphenols, mycoestrogens) which are hER panagonists and hERβ selective agonists displayed greater potency for zfERα as compared to zfERβs. Among hERα selective synthetic agonists, PPT did not activate zfERα while 16α-LE2 was the most zfERα selective compound. Altogether, these results confirm that all hER ligands control in a similar manner the transcriptional activity of zfERs although significant differences in selectivity were observed among subtypes. The zfER subtype selective ligands that we identified thus represent new valuable tools to dissect the physiological roles of the different zfERs. Finally, our work also points out that care has to be taken in transposing the results obtained using the zebrafish as a model for human physiopathology.
Product Description
Estrone, European Pharmacopoeia (EP) Reference Standard
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Estrone, ≥99%
Supelco
Genistein, analytical standard
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
4-tert-Octylphenol, 97%
Supelco
4-tert-Octylphenol, analytical standard
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
2,2′,4,4′-Tetrahydroxybenzophenone, 97%
Supelco
4-Hydroxytamoxifen, (E) and (Z) isomers (50:50), analytical standard
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Supelco
4-tert-Octylphenol solution, 1000 μg/mL in acetone, analytical standard