MilliporeSigma
  • Gene transfer properties and structural modeling of human stem cell-derived AAV.
Gene transfer properties and structural modeling of human stem cell-derived AAV.
Molecular therapy : the journal of the American Society of Gene Therapy (2014-06-14)
Laura J Smith, Taihra Ul-Hasan, Sarah K Carvaines, Kim Van Vliet, Ethel Yang, Kamehameha K Wong, Mavis Agbandje-McKenna, Saswati Chatterjee
ABSTRACT
Adeno-associated virus (AAV) vectors are proving to be remarkably successful for in vivo gene delivery. Based upon reports of abundant AAV in the human marrow, we tested CD34(+) hematopoietic stem cells for the presence of natural AAV. Here, we report for the first time, the presence of novel AAV variants in healthy CD34(+) human peripheral blood stem cells. The majority of healthy peripheral blood stem cell donors were found to harbor AAV in their CD34(+) cells. Every AAV isolated from CD34(+) cells mapped to AAV Clade F. Gene transfer vectors derived from these novel AAVs efficiently underwent entry and postentry processing in human cord blood stem cells and supported stable gene transfer into long-term, in vivo engrafting human HSCs significantly better than other serotypes. AAVHSC-transduced human CD34(+) cells engrafted in vivo and gave rise to differentiated transgene-expressing progeny. Importantly, gene-marked CD34(+) stem cells persisted long term in xenograft recipients, indicating transduction of primitive progenitors. Notably, correlation of structure with function permitted identification of potential capsid components important for HSC transduction. Thus, AAVHSCs represent a new class of genetic vectors for the manipulation of HSC genomes.
Product Description
Supelco
Sulfamethoxazole
Supelco
Sulfamethoxazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfamethoxazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Sulfamethoxazole, European Pharmacopoeia (EP) Reference Standard
USP
Sulfamethoxazole, United States Pharmacopeia (USP) Reference Standard