MilliporeSigma
  • Antiepileptogenic effects of the selective COX-2 inhibitor etoricoxib, on the development of spontaneous absence seizures in WAG/Rij rats.
Antiepileptogenic effects of the selective COX-2 inhibitor etoricoxib, on the development of spontaneous absence seizures in WAG/Rij rats.
Brain research bulletin (2015-02-24)
Rita Citraro, Antonio Leo, Rosario Marra, Giovambattista De Sarro, Emilio Russo
ABSTRACT
Different data suggest the involvement of specific inflammatory pathways in the pathogenesis of epilepsy. Cyclooxygenase (COX), which catalyses the production of pro-inflammatory prostaglandins, may play a significant role in seizure-induced neuroinflammation and neuronal hyperexcitability. COX-2 is constitutively expressed in the brain and also increased during/after seizures. COX-2 inhibitors may thus attenuate inflammation associated with brain disorders. We studied whether early long-term treatment (17 consecutive weeks starting from 45 days postnatal age) with the non-steroidal anti-inflammatory drug etoricoxib (10 mg/kg/day per os), a selective COX-2 inhibitor, was able to prevent/reduce the development of absence seizures in WAG/Rij rats, a recognized animal model of absence epilepsy and epileptogenesis. Drug effects on the incidence, duration and properties of absence seizure spike-wave discharges (SWDs) were measured both 1 and 5 months after treatment withdrawal; furthermore, the acute effects of etoricoxib on SWDs in 6-month-old WAG/Rij rats were measured. Early long-term treatment (ELTT) with etoricoxib led to an ∼40% long-lasting (5 months) reduction in the development of spontaneous absence seizures in adult WAG/Rij rats thus exhibiting antiepileptogenic effects. Acutely administered etoricoxib (10 and 20mg/kg i.p.) also had anti-absence properties, significantly reducing the number and duration of SWDs by ∼50%. These results confirm the antiepileptogenic effects of COX-2 inhibitors and suggest the possible role of COX-2, prostaglandin synthesis and consequent neuroinflammation in the epileptogenic process underlying the development of absence seizures in WAG/Rij rats.
Product Description
Supelco
Etoricoxib, VETRANAL®, analytical standard