MilliporeSigma
  • Association of higher erythropoiesis stimulating agent dose and mortality in children on dialysis.
Association of higher erythropoiesis stimulating agent dose and mortality in children on dialysis.
Pediatric nephrology (Berlin, Germany) (2014-05-06)
Rachel M Lestz, Barbara A Fivush, Meredith A Atkinson
ABSTRACT
Higher doses of erythropoiesis-stimulating agents (ESA) have been associated with an increased risk of adverse outcomes in adults with chronic kidney disease (CKD) and end-stage kidney disease (ESRD), but to our knowledge no trials have been performed in children. We examined the association between ESA dose and all-cause mortality in a prevalent pediatric dialysis population. Retrospective cohort study utilizing national data on all prevalent dialysis patients aged <18 years from the Centers for Medicare and Medicaid Services' 2005 ESRD Clinical Performance Measures (CPM) project, linked to 18-month mortality records from the United States Renal Data System. Multivariate Cox proportional hazards regression was performed to determine the risk of mortality by mean weekly ESA dose. Eight-hundred and twenty-nine children were included in the analysis; 7 % died during follow-up. A higher proportion of patients receiving ESA doses in the highest category (erythropoietin ≥350 units/kg/week or darbepoetin ≥1.5 units/kg/week) died (50 % vs 28 %, p = 0.002), and also demonstrated a trend toward lower hemoglobin (11.0 vs 11.4 g/dL, p = 0.05). In multivariate analysis, patients receiving the highest dose of ESA demonstrated an increased risk of mortality (hazard ratio 3.37; p value <0.01). Higher ESA dose is independently associated with mortality in children on chronic dialysis.
Sigma-Aldrich
Erythropoietin human, EPO, recombinant, expressed in HEK 293 cells, suitable for cell culture