MilliporeSigma
  • Effects of an immunomodulatory therapy and chondroitinase after spinal cord hemisection injury.
Effects of an immunomodulatory therapy and chondroitinase after spinal cord hemisection injury.
Neurosurgery (2014-05-30)
Matthew J Grosso, Virgilio Matheus, Megan Clark, Nico van Rooijen, Christopher A Iannotti, Michael P Steinmetz
ABSTRACT
Individually, immunomodulatory therapy and chondroitinases have demonstrated neuroprotective and potential neuroregenerative effects following spinal cord injury. To investigate the therapeutic potential of combined immunomodulatory and chondroitin sulfate-glycosaminoglycan degradation therapy in spinal cord injury. A combined immunomodulatory treatment using (1) liposome-encapsulated clodronate (selectively depletes peripheral macrophages), and (2) rolipram (a selective type 4 phosphodiesterase inhibitor), along with the chondroitin sulfate proteoglycan-glycosaminoglycan-degrading enzyme, chondroitinase ABC (ChABC), was assessed for its potential to promote axonal regrowth and improve locomotor recovery following midthoracic spinal cord hemisection injury in adult rats. We demonstrate that combined treatment with liposomal clodronate, rolipram, and ChABC attenuates macrophage accumulation at the site of injury, reduces axonal die-back of injured dorsal column axons, and produces the greatest improvement in locomotor recovery at 6 weeks postinjury compared with controls and noncombined therapy. Anterograde and retrograde tracing revealed that delivery of clodronate, rolipram, and ChABC did not promote substantial axonal regeneration through the site of injury, although the treatment did limit the extent of axonal die-back. Histological assessments revealed that combined treatment with clodronate/rolipram and/or ChABC resulted in a significant reduction in lesion size and cystic cavitation in comparison with injured controls. Combined clodronate, rolipram, and ChABC treatment reduced the accumulation of macrophages within the injured spinal cord 7 weeks after injury. The present data suggest that delivery of an immunomodulatory therapy consisting of clodronate and rolipram, in combination with ChABC, reduces axonal injury and enhances neuroprotection, plasticity, and hindlimb functional recovery after hemisection spinal cord injury in adult rats.
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Sigma-Aldrich
Anti-CS antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution