MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The effect of breed and sex on sulfamethazine, enrofloxacin, fenbendazole and flunixin meglumine pharmacokinetic parameters in swine.
The effect of breed and sex on sulfamethazine, enrofloxacin, fenbendazole and flunixin meglumine pharmacokinetic parameters in swine.
Journal of veterinary pharmacology and therapeutics (2014-04-16)
J T Howard, R E Baynes, J D Brooks, J L Yeatts, B Bellis, M S Ashwell, P Routh, A T O'Nan, C Maltecca
ABSTRACT
Drug use in livestock has received increased attention due to welfare concerns and food safety. Characterizing heterogeneity in the way swine populations respond to drugs could allow for group-specific dose or drug recommendations. Our objective was to determine whether drug clearance differs across genetic backgrounds and sex for sulfamethazine, enrofloxacin, fenbendazole and flunixin meglumine. Two sires from each of four breeds were mated to a common sow population. The nursery pigs generated (n = 114) were utilized in a random crossover design. Drugs were administered intravenously and blood collected a minimum of 10 times over 48 h. A non-compartmental analysis of drug and metabolite plasma concentration vs. time profiles was performed. Within-drug and metabolite analysis of pharmacokinetic parameters included fixed effects of drug administration date, sex and breed of sire. Breed differences existed for flunixin meglumine (P-value<0.05; Cl, Vdss ) and oxfendazole (P-value<0.05, AUC0→∞ ). Sex differences existed for oxfendazole (P-value < 0.05; Tmax ) and sulfamethazine (P-value < 0.05, Cl). Differences in drug clearance were seen, and future work will determine the degree of additive genetic variation utilizing a larger population.
Product Description
Flunixin meglumine, European Pharmacopoeia (EP) Reference Standard
Supelco
Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
Meglumine, European Pharmacopoeia (EP) Reference Standard
Supelco
Flunixin Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Flunixin meglumine, ≥98% (HPLC)
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Enrofloxacin, United States Pharmacopeia (USP) Reference Standard
Enrofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Enrofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Supelco
Flunixin, VETRANAL®, analytical standard
Sigma-Aldrich
N-Methyl-D-glucamine, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)