MilliporeSigma
  • Effects of low-intensity pulsed ultrasound on the expression of cyclooxygenase-2 in mandibular condylar chondrocytes.
Effects of low-intensity pulsed ultrasound on the expression of cyclooxygenase-2 in mandibular condylar chondrocytes.
Journal of oral & facial pain and headache (2014-07-30)
Yasunori Iwabuchi, Kotaro Tanimoto, Yuki Tanne, Toshihiro Inubushi, Takashi Kamiya, Ryo Kunimatsu, Naoto Hirose, Tomomi Mitsuyoshi, Shaoching Su, Eiji Tanaka, Kazuo Tanne
ABSTRACT
To determine the effect of low-intensity pulsed ultrasound (LIPUS) on cyclooxygenase-2 (COX-2) expression and related mechanisms by using cultured articular chondrocytes derived from porcine mandibular condyles after treatment with interleukin-1 beta (IL-1β). Chondrocytes were derived from porcine mandibular condylar cartilage and cultured. The cells were treated with or without 10 ng/mL IL-1β. At the same time, the cells were exposed to LIPUS for 20 minutes. After LIPUS exposure, the conditioned medium was changed to a fresh one without IL-1β, and the cells were incubated for 0 to 24 hours. The effects of LIPUS on IL-1β-treated chondrocytes were examined in terms of the expression of p-integrin β1, COX-2, and phosphorylated extracellular signal-related kinase (p-ERK) 1/2 by real-time polymerase chain reaction (PCR) and Western blot analyses. Differences in the means among multiple groups were examined by one-way analysis of variance (ANOVA) for all groups at each time point, followed by a Scheffé multiple comparison test as a post-hoc test; Student t test was also used. COX-2 mRNA level was upregulated by the treatment with IL-1β and was suppressed significantly (P < .01) by LIPUS exposure. Furthermore, LIPUS enhanced gene expression and phosphorylation of integrin β, and it inhibited the expression of p-ERK 1/2. LIPUS exposure inhibited IL-1β-induced COX-2 expression through the integrin β1 receptor followed by the phosphorylation of ERK 1/2. Despite the restricted duration of its effect, LIPUS is suggested to be a potential candidate as a preventive and auxiliary treatment to suppress the degradation of articular chondrocytes in temporomandibular joint osteoarthritis.
Sigma-Aldrich
IL-1 beta human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture