MilliporeSigma
  • Randomized trial of peginterferon alpha-2b plus low and escalating dose of ribavirin in patients with chronic hepatitis C with high viral load genotype 1.
Randomized trial of peginterferon alpha-2b plus low and escalating dose of ribavirin in patients with chronic hepatitis C with high viral load genotype 1.
Journal of medical virology (2015-01-23)
Yoshiaki Iwasaki, Yasuyuki Araki, Hideaki Taniguchi, Ryo-Ichi Okamoto, Haruhiko Kobashi, Fusao Ikeda, Yasuhiro Miyake, Akinobu Takaki, Kazuhide Yamamoto
ABSTRACT
It has been reported that elderly patients with chronic hepatitis C infection cannot tolerate standard combination therapy. In this randomized, controlled trial, the efficacy and safety of peginterferon alpha-2b plus a low and escalating dose of ribavirin in chronic hepatitis C patients with high viral load genotype 1 were investigated. Sixty-two patients were randomized into combination therapy with standard ribavirin dosing (group 1) or low and escalating ribavirin dosing (group 2). Patients were evaluated for safety and efficacy of treatment. There was no significant difference in the prevalence of virological response between the groups throughout the treatment as well as 24 weeks after treatment. However, the response in patients ≥60 years of age was higher in group 1 than in group 2 at early treatment phase (P = 0.015). The prevalence of completion of therapy in patients ≥60 years of age tended to be higher in group 2 than in group 1 (50% vs. 0%, P = 0.055). There was no significant difference in dose modification of peginterferon alpha-2b between the groups. However, dose modification of ribavirin was significantly more frequent in group 1 than in group 2 (60% vs. 24%, P = 0.005). These data suggest that combination therapy with low and escalating dosing of ribavirin may be safer in elderly patients than that with standard dosing of ribavirin without impairing the treatment response.
Sigma-Aldrich
Ribavirin, antiviral
Ribavirin, European Pharmacopoeia (EP) Reference Standard