MilliporeSigma
  • Multiscale cardiac modelling reveals the origins of notched T waves in long QT syndrome type 2.
Multiscale cardiac modelling reveals the origins of notched T waves in long QT syndrome type 2.
Nature communications (2014-09-26)
Arash Sadrieh, Luke Domanski, Joe Pitt-Francis, Stefan A Mann, Emily C Hodkinson, Chai-Ann Ng, Matthew D Perry, John A Taylor, David Gavaghan, Rajesh N Subbiah, Jamie I Vandenberg, Adam P Hill
ABSTRACT
The heart rhythm disorder long QT syndrome (LQTS) can result in sudden death in the young or remain asymptomatic into adulthood. The features of the surface electrocardiogram (ECG), a measure of the electrical activity of the heart, can be equally variable in LQTS patients, posing well-described diagnostic dilemmas. Here we report a correlation between QT interval prolongation and T-wave notching in LQTS2 patients and use a novel computational framework to investigate how individual ionic currents, as well as cellular and tissue level factors, contribute to notched T waves. Furthermore, we show that variable expressivity of ECG features observed in LQTS2 patients can be explained by as little as 20% variation in the levels of ionic conductances that contribute to repolarization reserve. This has significant implications for interpretation of whole-genome sequencing data and underlies the importance of interpreting the entire molecular signature of disease in any given individual.