MilliporeSigma
  • Advanced gynecologic malignancies treated with a combination of the VEGF inhibitor bevacizumab and the mTOR inhibitor temsirolimus.
Advanced gynecologic malignancies treated with a combination of the VEGF inhibitor bevacizumab and the mTOR inhibitor temsirolimus.
Oncotarget (2014-04-20)
Sarina A Piha-Paul, Jennifer J Wheler, Siqing Fu, Charles Levenback, Karen Lu, Gerald S Falchook, Aung Naing, David S Hong, Apostolia M Tsimberidou, Razelle Kurzrock
ABSTRACT
Bevacizumab and temsirolimus are active agents in gynecologic tumors. Temsirolimus attenuates upregulation of HIF-1α levels, a resistance mechanism for antiangiogenics, and targets the PI3-kinase/AKT/mTOR axis, commonly aberrant in these tumors. We analyzed safety and responses in 41 patients with gynecologic cancers treated as part of a Phase I study of bevacizumab and temsirolimus. Median age of the 41 women was 60 years (range, 33-80 years); median number of prior systemic therapies was 4 (1-11). Grade 3 or 4 treatment-related toxicities included: thrombocytopenia (10%), mucositis (2%), hypertension (2%), hypercholesterolemia (2%), fatigue (7%), elevated aspartate aminotransferase (2%), and neutropenia (2%). Twenty-nine patients (71%) experienced no treatment-related toxicity greater than grade 2. Full FDA-approved doses of both drugs (bevacizumab 15mg/kg IV Q3weeks and temsirolimus 25mg IV weekly) were administered without dose-limiting toxicity. Eight patients (20%) achieved stable disease (SD) > 6 months and 7 patients (17%), a partial response (PR) [total = 15/41 patients (37%)]. Eight of 13 patients (62%) with high-grade serous histology (ovarian or primary peritoneal) achieved SD > 6 months/PR. Bevacizumab and temsirolimus was well tolerated. Thirty-seven percent of heavily-pretreated patients achieved SD > 6 months/PR, suggesting that this combination warrants further study.