MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Comparison of image quality of shoulder CT arthrography conducted using 120 kVp and 140 kVp protocols.
Comparison of image quality of shoulder CT arthrography conducted using 120 kVp and 140 kVp protocols.
Korean journal of radiology (2014-12-04)
Se Jin Ahn, Sung Hwan Hong, Jee Won Chai, Ja-Young Choi, Hye Jin Yoo, Sae Hoon Kim, Heung Sik Kang
ABSTRACT
To compare the image quality of shoulder CT arthrography performed using 120 kVp and 140 kVp protocols. Fifty-four CT examinations were prospectively included. CT scans were performed on each patient at 120 kVp and 140 kVp; other scanning parameters were kept constant. Image qualities were qualitatively and quantitatively compared with respect to noise, contrast, and diagnostic acceptability. Diagnostic acceptabilities were graded using a one to five scale as follows: 1, suboptimal; 2, below average; 3, acceptable; 4, above average; and 5, superior. Radiation doses were also compared. Contrast was better at 120 kVp, but noise was greater. No significant differences were observed between the 120 kVp and 140 kVp protocols in terms of diagnostic acceptability, signal-to-noise ratio, or contrast-to-noise ratio. Lowering tube voltage from 140 kVp to 120 kVp reduced the radiation dose by 33%. The use of 120 kVp during shoulder CT arthrography reduces radiation dose versus 140 kVp without significant loss of image quality.
Sigma-Aldrich
N-Methyl-D-glucamine, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)
Supelco
Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
Meglumine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications