MilliporeSigma
  • Ubiquitin-specific protease 2a stabilizes MDM4 and facilitates the p53-mediated intrinsic apoptotic pathway in glioblastoma.
Ubiquitin-specific protease 2a stabilizes MDM4 and facilitates the p53-mediated intrinsic apoptotic pathway in glioblastoma.
Carcinogenesis (2014-01-22)
Chun-Lin Wang, Jun-Yu Wang, Zhen-Yang Liu, Xiao-Mei Ma, Xiao-Wen Wang, Hai Jin, Xiao-Ping Zhang, Da Fu, Li-Jun Hou, Yi-Cheng Lu
ABSTRACT
The mouse double minute 4 (MDM4) oncoprotein may inhibit tumorigenesis by regulating the apoptotic mediator p53. Ubiquitin-specific protease 2a (USP2a) is a deubiquitinating enzyme that protects MDM4 against degradation, so USP2-MDM4 interaction may be a key determinant of the malignant potential of human cancers. MDM4 and USP2a, as well as the MDM4-USP2a complex, were more highly expressed in glioblastoma multiforme tissue samples from patients with good prognosis compared with patients with poor prognosis. Analysis of the prognostic parameters indicated that MDM4 expression was positively correlated with an increased likelihood for survival. Compared with the poor prognosis patients, mitochondria from good prognosis glioma patients contained higher levels of both MDM4 and the proapoptotic protein p53Ser46(P). In U87MG glioma cell line, the overexpression of MDM4 enhanced ultraviolet (UV)-induced cytochrome c release and apoptosis. In contrast, MDM4 knockdown decreased mitochondrial p53Ser46(P) levels and rescued cells from UV-induced apoptosis. The expression of MDM4 and USP2a were positively correlated with each other. MDM4-USP2a complexes were found only in the cytoplasmic fraction, whereas the mitochondrial fraction contained MDM4-p53Ser46(P) and MDM4-Bcl-2 complexes. Overexpression of USP2a increased p53 and p53Ser46(P) levels in the mitochondria, whereas simultaneous MDM4 knockdown completely reversed this effect. UV-induced apoptosis was reduced by USP2a knockdown but restored by the simultaneous overexpression of MDM4. This apoptotic response was reduced by knockdown of p53 but not p21. Our results suggest that USP2a binds to and stabilizes MDM4; thus in turn, it enhances the mitochondrial localization of p53 and promotes apoptosis in glioma cells.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone AA13, purified from hybridoma cell culture