MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • BODIPY(®) FL histamine as a new modality for quantitative detection of histamine receptor upregulation upon IgE sensitization in murine bone marrow-derived mast cells.
BODIPY(®) FL histamine as a new modality for quantitative detection of histamine receptor upregulation upon IgE sensitization in murine bone marrow-derived mast cells.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2014-09-13)
Arash Mirzahosseini, Marianna Kovács, Károly Kánai, Péter Csutora, Balázs Dalmadi
ABSTRACT
Flow cytometry is one of the most widely used methods for the qualitative and quantitative analysis of cell surface expressed proteins by making use of fluorescent specific antibodies. Lacking an antibody validated for flow cytometry, an alternative approach for labeling cell surface receptors is the use of fluorescently tagged ligands. In this study, histamine H4 receptor transfected Chinese hamster ovary cells and murine bone marrow-derived mast cells (mBMMCs) were selected for studying the possibility of staining individual histamine receptors using BODIPY(®) FL histamine and selective antagonists. Flow cytometric measurements and supporting calculations showed that BODIPY FL histamine is suitable tool for quantitating cell surface histamine receptors. The binding, and competitive inhibition of this fluorescent ligand were characterized, which were in good agreement with a semi-empirical model constructed from fundamental protein-binding relationships. Using this method it was shown for the first time that even though mature mBMMCs express H2R and H4R to the same extent, immunoglobulin E sensitization results in H4R upregulation only, while the surface expression of H2R remains unchanged.
Product Description
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
Cimetidine, European Pharmacopoeia (EP) Reference Standard
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
USP
Cimetidine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Cimetidine
Sigma-Aldrich
Streptomycin sulfate salt, powder
Supelco
Histamine, analytical standard
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Cimetidine for peak identification, European Pharmacopoeia (EP) Reference Standard
Cimetidine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard