MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Identification of proteins associated with lymph node metastasis of gastric cancer.
Identification of proteins associated with lymph node metastasis of gastric cancer.
Journal of cancer research and clinical oncology (2014-05-16)
Yan Ma, Yan-Feng Li, Tie Wang, Rui Pang, Ying-Wei Xue, Shu-Peng Zhao
ABSTRACT
The aim of this study was to identify proteins associated with gastric cancer lymph node metastasis and explore the clinicopathological significance of these proteins. Gastric cancer tissues were obtained from 24 patients with high or low lymph node metastatic potential. Total cellular proteins were separated by two-dimensional gel electrophoresis (2-DE), analyzed by MALDI/TOF-TOF MS, and identified by a database search. Expression of 14-3-3β and profilin-1 was then immunohistochemically verified in paraffin-embedded gastric cancer tissues from 128 patients and analyzed by multivariate logistic regression models, Kaplan-Meier curves, and Cox proportional hazard models. A total of 26 differentially expressed proteins were identified, 20 of which were overexpressed and 6 of which were underexpressed. 14-3-3β and profilin-1 were upregulated in gastric cancer tissues with and without lymph node metastasis, respectively. Expression of 14-3-3β protein was associated, but profilin-1 expression was inversely associated with gastric cancer lymph node metastasis. Multivariate analysis showed that overexpression of 14-3-3β and reduced expression of profilin-1 were independent risk factors for gastric cancer lymph node metastasis, while 14-3-3β overexpression was an independent prognostic factor for gastric cancer patients. The current study identified 26 differentially expressed proteins. Further studies showed that both 14-3-3β and profilin-1 protein may be useful biomarkers for prediction of gastric cancer lymph node metastasis and that expression of 14-3-3β was a prognostic marker for gastric cancer patients.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, Ultra pure
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder