MilliporeSigma
  • The differential antiemetic properties of GLP-1 receptor antagonist, exendin (9-39) in Suncus murinus (house musk shrew).
The differential antiemetic properties of GLP-1 receptor antagonist, exendin (9-39) in Suncus murinus (house musk shrew).
Neuropharmacology (2014-04-15)
Sze Wa Chan, Zengbing Lu, Ge Lin, David Tai Wai Yew, Chi Kong Yeung, John A Rudd
ABSTRACT
The use of glucagon-like peptide-1 (7-36) amide (GLP-1) receptor agonists for the treatment of type 2 diabetes mellitus is commonly associated with nausea and vomiting. Previous studies using Suncus murinus revealed that the GLP-1 receptor agonist, exendin-4, induces emesis via the brainstem and/or hypothalamus. The present study investigated the mechanism of exendin-4-induced emesis in more detail. Ondansetron (1 mg/kg, s.c.) and CP-99,994 (10 mg/kg, s.c) failed to reduce emesis induced by exendin-4 (3 nmol, i.c.v.), suggesting that 5-HT3 and NK1 receptors are not involved in the mechanism. In other studies, the GLP-1 receptor antagonist, exendin (9-39), antagonised emesis and c-Fos expression in the brainstem and the paraventricular hypothalamus induced by the chemotherapeutic drug cisplatin (30 mg/kg, i.p.; p < 0.05), but not the emesis induced by nicotine (5 mg/kg, s.c.; p > 0.05), or copper sulphate pentahydrate (120 mg/kg, p.o.; p > 0.05). GLP-1 receptors may therefore represent a potential target for drugs to prevent chemotherapy-induced emesis in situations where 5-HT3 and NK1 receptor antagonists fail.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard