MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Visceral white adipose tissue is susceptible to alcohol-induced lipodystrophy in rats: role of acetaldehyde.
Visceral white adipose tissue is susceptible to alcohol-induced lipodystrophy in rats: role of acetaldehyde.
Alcoholism, clinical and experimental research (2015-02-24)
Wenliang Zhang, Wei Zhong, Xiuhua Sun, Qian Sun, Xiaobing Tan, Qiong Li, Xinguo Sun, Zhanxiang Zhou
ABSTRACT
Chronic alcohol exposure causes lipid dyshomeostasis at the adipose-liver axis, reducing lipid storage in white fat and increasing lipid deposit in the liver. Previous studies have shown that visceral fat, rather than subcutaneous fat, is a risk factor for metabolic diseases. This study was conducted to determine whether chronic alcohol exposure differentially affects lipid metabolism in visceral (epididymal) and subcutaneous fat, and the mechanisms underlying the alcohol effects. Male Wistar rats were pair-fed the Lieber-DeCarli control or alcohol liquid diet for 12 weeks to determine the effects of alcohol on the white fat. Tissue explants culture and 3T3-L1 culture were conducted to define the role of acetaldehyde in alcohol-induced adipose tissue dysfunction. Chronic alcohol feeding significantly reduced visceral fat mass and down-regulated peroxisome proliferator activator receptor-γ and CCAAT/enhancer binding protein-α, 2 important transcription factors in regulation of lipogenesis. The protein levels of lipogenic enzymes including phospho-ATP-citrate lyase, acetyl-CoA carboxylase, fatty acid synthase, lipin1, and diacylglycerol acyltransferase 2 in the visceral fat were reduced. In contrast, chronic alcohol exposure did not affect subcutaneous fat mass and had less effect on the protein levels of lipogenic enzymes and regulators. Accordingly, the visceral fat showed a lower protein level of aldehyde detoxification enzymes compared to the subcutaneous fat. Acetaldehyde treatment to either visceral fat explants or 3T3-L1 adipocytes produced similar effects on lipogenic enzymes and regulators as observed in animal model. These results demonstrated that visceral fat is more susceptible to alcohol toxicity compared to subcutaneous fat, and disruption of adipose lipogenesis contributes to the pathogenesis of alcoholic lipodystrophy.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder