MilliporeSigma
  • Computational simulation and statistical analysis on the relationship between corrosion inhibition efficiency and molecular structure of some phenanthroline derivatives on mild steel surface.
Computational simulation and statistical analysis on the relationship between corrosion inhibition efficiency and molecular structure of some phenanthroline derivatives on mild steel surface.
International Journal of Electrochemical Science (2011)
Obi-Egbedi NO, et al.