MilliporeSigma
  • A convenient synthesis of 2,4-diarylpolyhydroquinoline derivations in the presence of ammonium acetate.
A convenient synthesis of 2,4-diarylpolyhydroquinoline derivations in the presence of ammonium acetate.
Synthetic Communications (2002)
Wang XS, et al.