MilliporeSigma
  • A general method for the Suzuki-Miyaura cross-coupling of sterically hindered aryl chlorides: Synthesis of di-and tri-ortho-substituted biaryls in 2-propanol at room temperature.
A general method for the Suzuki-Miyaura cross-coupling of sterically hindered aryl chlorides: Synthesis of di-and tri-ortho-substituted biaryls in 2-propanol at room temperature.
Journal of the American Chemical Society (2003)
Navarro O, et al.