MilliporeSigma
  • (R)-PFI-2 is a potent and selective inhibitor of SETD7 methyltransferase activity in cells.
(R)-PFI-2 is a potent and selective inhibitor of SETD7 methyltransferase activity in cells.
Proceedings of the National Academy of Sciences of the United States of America (2014-08-20)
Dalia Barsyte-Lovejoy, Fengling Li, Menno J Oudhoff, John H Tatlock, Aiping Dong, Hong Zeng, Hong Wu, Spencer A Freeman, Matthieu Schapira, Guillermo A Senisterra, Ekaterina Kuznetsova, Richard Marcellus, Abdellah Allali-Hassani, Steven Kennedy, Jean-Philippe Lambert, Amber L Couzens, Ahmed Aman, Anne-Claude Gingras, Rima Al-Awar, Paul V Fish, Brian S Gerstenberger, Lee Roberts, Caroline L Benn, Rachel L Grimley, Mitchell J S Braam, Fabio M V Rossi, Marius Sudol, Peter J Brown, Mark E Bunnage, Dafydd R Owen, Colby Zaph, Masoud Vedadi, Cheryl H Arrowsmith
ABSTRACT
SET domain containing (lysine methyltransferase) 7 (SETD7) is implicated in multiple signaling and disease related pathways with a broad diversity of reported substrates. Here, we report the discovery of (R)-PFI-2-a first-in-class, potent (Ki (app) = 0.33 nM), selective, and cell-active inhibitor of the methyltransferase activity of human SETD7-and its 500-fold less active enantiomer, (S)-PFI-2. (R)-PFI-2 exhibits an unusual cofactor-dependent and substrate-competitive inhibitory mechanism by occupying the substrate peptide binding groove of SETD7, including the catalytic lysine-binding channel, and by making direct contact with the donor methyl group of the cofactor, S-adenosylmethionine. Chemoproteomics experiments using a biotinylated derivative of (R)-PFI-2 demonstrated dose-dependent competition for binding to endogenous SETD7 in MCF7 cells pretreated with (R)-PFI-2. In murine embryonic fibroblasts, (R)-PFI-2 treatment phenocopied the effects of Setd7 deficiency on Hippo pathway signaling, via modulation of the transcriptional coactivator Yes-associated protein (YAP) and regulation of YAP target genes. In confluent MCF7 cells, (R)-PFI-2 rapidly altered YAP localization, suggesting continuous and dynamic regulation of YAP by the methyltransferase activity of SETD7. These data establish (R)-PFI-2 and related compounds as a valuable tool-kit for the study of the diverse roles of SETD7 in cells and further validate protein methyltransferases as a druggable target class.