MilliporeSigma
  • Grb2 interacts with SGEF and antagonizes the ability of SGEF to enhance EGF-induced ERK1/2 activation.
Grb2 interacts with SGEF and antagonizes the ability of SGEF to enhance EGF-induced ERK1/2 activation.
Molecular and cellular biochemistry (2014-01-09)
Hongtao Wang, Shanhu Li, Hailiang Li, Peng Wang, Fang Huang, Yali Zhao, Lan Yu, Guolan Luo, Xiaoqing Zhang, Jian Wang, Jianguang Zhou
ABSTRACT
Previously, we demonstrated that SGEF enhances EGFR stability; however, SGEF-mediated downstream signaling of EGFR is not well understood. Here, we show that SGEF enhances EGF-induced ERK1/2 activation independent of its guanine nucleotide exchange (GEF) activity. We further show that SGEF interacts with Grb2, a critical downstream transducer of EGFR. Surprisingly, we found that interaction of Grb2 to SGEF antagonizes the ability of SGEF to enhance EGF-induced ERK1/2 activation. Taken together, this study reports a novel function of SGEF that excludes GEF and also provides important insights into the complex role of Grb2 in EGFR signal transduction.