MilliporeSigma
  • Ablation of uroplakin III gene results in small urothelial plaques, urothelial leakage, and vesicoureteral reflux.
Ablation of uroplakin III gene results in small urothelial plaques, urothelial leakage, and vesicoureteral reflux.
The Journal of Cell Biology (2000)
Hu P