MilliporeSigma
  • Magnesia-supported hydroxylamine hydrochloride in the presence of sodium carbonate as an efficient reagent for the synthesis of 1, 2, 4-oxadiazoles from nitriles.
Magnesia-supported hydroxylamine hydrochloride in the presence of sodium carbonate as an efficient reagent for the synthesis of 1, 2, 4-oxadiazoles from nitriles.
Tetrahedron Letters (2007)
Kaboudin B and Saadati F.